Fisherbrand™ Hemostats Rochester-Ochsner Forceps
Fisherbrand Hemostats Rochester-Ochsner (Kocher) are primarily used to handle to aggressively handle and manipulate of thick tissue or clamp heavy or dense vessels. ASTM™ standard (F899-84) stainless steel.
 View more versions of this product
Catalog No. 13-812-18
Explore available promotions
Description
Description
  • Fisherbrand™ Hemostats Rochester-Ochsner (Kocher) Forceps are primarily used to handle to aggressively handle and manipulate of thick tissue or clamp heavy or dense vessels.The jaws have serrated inner lining with 1 x 2 teeth at the tipf for secured tissue grasping
  • Our forceps are designed to exceed perfromance expectations -This is the Fisherbrand Advantage.
  • 3-Stage Locking System – Variable blood flow control (Stage 0: complete flow to Stage 3: no flow)
  • Finger-ring handle designed for comfort, stability and ease of use
  • Multipurpose - Useful in educational, research or healthcare setting
  • Box-lock joint for precision clamping
  • Hand polished to mirror finish
  • Made from surgical stainless steel
  • Electronically passivated for corrosion resistance
  • Can be sterilized by Eto/gamma radiation
  • Rockwell hardness (HRC): 40-45
  • Autoclavable up to 135°C
  • Latex-free
Specifications
Specifications
Rochester Ochsner Forcep
ASTM, ISO 9001, cGMP
Serrated jaws, box lock joint
Straight
18.4 cm
Autoclavable
Stainless Steel
1 x 2
7.25 in.