Fisherbrand™ Histology Buckets
Flip-top lid helps to reduce noxious odors
Description
- Opaque polypropylene construction resists cracking and chemicals, including formaldehyde
- Metal carrying handle eases transport
- Easy to stack and store
- No graduation marks
- Autoclavable
Specifications
No | |
1.37 gal. | |
Flip-top Snap-on Lid |
Polypropylene | |
5 L |