Fisherbrand™ Histology Buckets
Flip-top lid helps to reduce noxious odors
 View more versions of this product
Catalog No. 23-111-252
Explore available promotions
Description
Description
  • Opaque polypropylene construction resists cracking and chemicals, including formaldehyde
  • Metal carrying handle eases transport
  • Easy to stack and store
  • No graduation marks
  • Autoclavable
Specifications
Specifications
No
2 gal.
Flip-top Snap-on Lid
Polypropylene
8 L