Fisherbrand™ Histosette™ II Biopsy Processing/Embedding Cassettes with Separate Lid and Base
Histosette™ II Biopsy Processing/Embedding Cassettes with Separate Lid and Base open and close as often as necessary - cassettes will always relock securely.
Supplier: Fisherbrand™ M4867FS
Description
- Disposable, plastic cassettes hold biopsy specimens during embedding, processing, and storage
- Molded from a special high-density polymer which is resistant to the chemical action of histological solvents
- Efficient flow-through slots maximize fluid exchange and ensure proper drainage
- The anterior writing surface is slanted at a 45° angle, making the cassette more suitable for use with certain types of labeling instruments
Order Info
Available in 11 noncytotoxic, nonmetallic colors.
Specifications
Slots | |
Separate Lid | |
Plastic |
45° | |
Peach | |
Biopsy Cassette |