missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ HMIG and NFR Dispenser Labels
Keep workers safe by properly informing them of workplace hazards.
$7.32 - $10.78
Specifications
Color | Assorted |
---|---|
Format | Label |
For Use With (Application) | Keep workers safe by properly informing them of workplace hazards |
Catalog Number | Mfr. No. | Product Type | Quantity | Size (English) | Size (Metric) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Quantity | Size (English) | Size (Metric) | Price | Quantity | ||
18-999-784
|
Fisherbrand™
775 |
HMIG Dispenser Label | 500/Pk. | 0.87 x 2.87 in. | 0.02 x 0.079 cm |
Each for $10.78
|
|
||
18-999-785
|
Fisherbrand™
67702 |
HMIG Dispenser Label | 750/Pk. | 0.87 x 0.87 in. | 0.02 x 02 cm |
Each for $7.32
|
|
||
18-999-786
|
N/A | NFR Dispenser Label | 500/Pk. | 0.87 x 2.87 in. | 0.02 x 0.079 cm |
Each for $7.32
|
|
||
18-999-787
|
N/A | NFR Dispenser Label | 750/Pk. | 0.87 x 0.87 in. | 0.02 x 02 cm |
Each for $7.32
|
|
||
Description
- Rolls come in a dispenser pack for convenient storage and use
- HMIG labels feature a write-on area for specific hazard information
- 18-999-786 NFR labels have room to write important information
- 18-999-787 labels feature an NFR legend: call attention to various chemical hazards throughout the workplace
Order Info
- Come in a dispenser pack for convenient storage and use
- Hazardous Materials Identification Guide labels come in packs of 500 and 750, feature a write-on area for specific hazard information
- 18-999-786 National Fire Rating labels come in packs of 500 and feature room to write important information
- 18-999-787 labels are available in packs of 750 and feature an NFR legend, calling attention to various chemical hazards throughout the workplace
Specifications
Assorted | |
Label | |
Keep workers safe by properly informing them of workplace hazards |