Fisherbrand™ Hygrometer Replacement Battery
Hygrometer Replacement Battery for use with Hygrometers.
$16.63 - $24.44
Catalog Number | Mfr. No. | Battery Type | Product Type | Voltage | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Battery Type | Product Type | Voltage | Price | Quantity | |||
02-440-1A
|
N/A | CR2032 | Lithium Coin Battery | 3 V |
Each for $24.44
|
|
|||
02-440-1C
|
N/A | 357 | Button Cell Battery | 1.5 V |
Each for $16.63
|
|
|||
02-440-1D
|
N/A | 392 | Button Cell Battery | 1.5 V |
Each for $16.88
|
|
|||
Description
Hygrometer Replacement Battery for use with Hygrometers.
- Come with lithium, button cell and “AAA” alkaline battery
- The electrical requirements are 3 V, 1.5 V and 1.5 V respectively
Specifications
For hygrometers |