Fisherbrand™ Infecon™ 2000 and 2050 Infectious/Diagnostic Shippers
For shipping test tubes containing infectious substances or diagnostic specimens
Description
For shipping test tubes containing infectious substances or diagnostic specimens
- Bubble-wrap compartment
- Liquid-tight, plastic secondary container withstands pressure up to 95kPa
- U.N. markings and a label for both infectious substances and diagnostic specimens
Compliance
Complies with ICAO/IATA Packing Instruction 602
Specifications
Complies with ICAO/IATA Packing Instruction 602 | |
For shipping test tubes containing infectious substances or diagnostic specimens | |
4 x 4 x 8 in. | |
Infecon 2000, Infectious/Diagnostic Shipper |