Fisherbrand™ InkJet Plus Microscope Slides
Superfrost™/ColorFrost™ microscope slides designed for use with Leica™ Microsystems IP S inkjet printer
 View more versions of this product
Catalog No. 12550111
Explore available promotions
Description
Description
Superfrost™/ColorFrost™ microscope slides designed for use with Leica™ Microsystems IP S inkjet printer
  • Printing instruments use chemical-resistant ink and print directly on Superfrost/ColorFrost tab of the slide
  • For histology, cytology, hematology, and microbiology applications
  • Clipped 45° or straight 90° corners
  • Include a plus (+) charge symbol
  • Frosted tabs in nine different colors
Specifications
Specifications
Tabs
Green
90°