Fisherbrand™ Sterilization Pouches Edge
Edge
No tape or heat sealing needed
Supplier:  Fisherbrand™ SCL1218
 View more versions of this product
Catalog No. 01-812-58
Edge
Explore available promotions
Description
Description
No tape or heat sealing needed
Integral sealing strip eliminates the need to tape or heat seal pouches. Simply peel off protective strip, fold along perforation and press firmly to seal pouch.
  • Triple heat-processed seams provide excellent tear and puncture resistance
  • Suitable for steam or EtO gas sterilization
  • Indicator arrows change color when sterilization cycle is complete
  • Made of paper/plastic; medical-grade paper side guards against puncture
  • 2mil-thick blue transparent polyester/polypropylene laminate film side allows contents to be identified quickly and shows any compromise in the film
  • Corner tack seals prevent contamination in the seal area
  • Thumb notch makes opening pouch easy
Specifications
Specifications
Blue
Non-sterile
18 x 12 in.
30 cm
46 cm
Paper, Plastic
2 mil
12 in.
18 in.