Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
Fisherbrand™ Institutional Trash Can Liners
Provide a leak resistant containment solution for non-medical waste disposal
$102.52 - $244.48
Spécifications
Product Type | Institutional Trash Can Liner |
---|---|
For Use With (Application) | Collecting and containing refuse, non medical waste disposal |
Avertissement:
Not for use with biohazardous waste.
Numéro de catalogue | Numéro du manufacturier. | Capacity (English) | Color | Material | Thickness (English) | Prix | Quantité | ||
---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Capacity (English) | Color | Material | Thickness (English) | Prix | Quantité | ||
14-828-401
|
N/A | 12 to 16 gal. | Clear | LLDPE | 0.35 mil |
caisse de 1000 chaque for $228.29
|
|
||
14-828-405
|
N/A | 32 to 33 gal. | Brown | LLDPE | 0.65 mil |
caisse de 250 chaque for $134.87
|
|
||
14-828-404
|
N/A | 32 to 33 gal. | Clear | LLDPE | 0.65 mil |
caisse de 250 chaque for $128.19
|
|
||
14-828-406
|
N/A | 32 to 33 gal. | Clear | LLDPE | 1.15 mil |
caisse de 100 chaque for $102.52
|
|
||
14-828-407
|
N/A | 40 to 45 gal. | Clear | LLDPE | 0.80 mil |
caisse de 250 chaque for $193.23
|
|
||
14-828-408
|
N/A | 40 to 45 gal. | Clear | LLDPE | 1.15 mil |
caisse de 100 chaque for $125.10
|
|
||
14-828-409
|
N/A | 60 gal. | Clear | LLDPE | 0.80 mil |
caisse de 200 chaque for $225.74
|
|
||
14-828-411
|
N/A | 56 gal. | Brown | LLDPE | 1.50 mil |
caisse de 100 chaque for $190.48
|
|
||
14-828-414
|
N/A | 20 to 30 gal. | Clear | HDPE | 0.40 mil |
caisse de 500 chaque for $215.94
|
|
||
14-828-417
|
N/A | 33 gal. | Clear | HDPE | 0.63 mil |
caisse de 250 chaque for $185.97
|
|
||
14-828-418
|
N/A | 40 to 45 gal. | Clear | HDPE | 0.47 mil |
caisse de 250 chaque for $199.10
|
|
||
14-828-419
|
N/A | 40 to 45 gal. | Clear | HDPE | 0.63 mil |
caisse de 250 chaque for $244.48
|
|
||
14-828-420
|
N/A | 60 gal. | Clear | HDPE | 0.63 mil |
caisse de 200 chaque for $207.29
|
|
||
Description
Provide a leak resistant containment solution for non-medical waste disposal
- For collecting and containing refuse
- Available in both linear low density (LLDPE) and high density (HDPE) polyethylene film
- Sold in flat packs for easy dispensing
- Provide a cost-effective solution for every application
Spécifications
Institutional Trash Can Liner | |
Collecting and containing refuse, non medical waste disposal |