Fisherbrand™ Kahn Serological Pipets
For measuring and transferring serum and antigen in Kahn tests.
Description
Used in Kahn tests as published in American Journal of Public Health, 129 (June 1924).
- Accuracy printed on each pipet
- Borosilicate glass with permanent amber markings
- Tuf-Temp™ treated to reduce chipping
Compliance
Calibrated To Deliver/Blow Out. Pipet 13-677D meets ASTM™ E 1044-96.
Specifications
±0.008 mL | |
Yes | |
Blue | |
Serological Pipet |
Borosilicate Glass | |
0.001 mL | |
0.2 mL |