Fisherbrand™ Kahn Serological Pipets
For measuring and transferring serum and antigen in Kahn tests.
 View more versions of this product
Catalog No. 13-677G
Explore available promotions
Description
Description
Used in Kahn tests as published in American Journal of Public Health, 129 (June 1924).
  • Accuracy printed on each pipet
  • Borosilicate glass with permanent amber markings
  • Tuf-Temp™ treated to reduce chipping
Compliance
Calibrated To Deliver/Blow Out. Pipet 13-677D meets ASTM™ E 1044-96.
Specifications
Specifications
±0.005 mL
Yes
Yellow
Serological Pipet
Borosilicate Glass
0.001 mL
0.1 mL