Fisherbrand™ Disposable Inoculating Loops and Needles, Flexible Needle and Loop
For quantitative procedures such as sampling, serial dilutions and urine counts, as well as for bacterial inoculation. Fisherbrand™ Disposable Inoculating Loops and Needles, Length: 9 in. (23cm) are molded from a specially formulated, biologically inert polymer.
 View more versions of this product
Catalog No. 22-363-598
Explore available promotions
Description
Description
For quantitative procedures such as sampling, serial dilutions and urine counts, as well as for bacterial inoculation. Fisherbrand™ Disposable Inoculating Loops and Needles, Length: 9 in. (23cm) are molded from a specially formulated, biologically inert polymer.
  • Flexible enough to facilitate streaking, yet rigid enough to penetrate the media for subsurface inoculation
  • Gamma-irradiated for sterility in accordance with U.S. Pharmacopoeia, Volume XX
  • Combi Loop has a 10μL loop on one end and a 1μL loop on the other
  • Color coded for easy identification
Specifications
Specifications
Single-use
Biologically Inert Polymer
Sterile
207 mm
Clear
Certificate of dimensional accuracy
Ziplock Bag
0.09 cm
8.15 in.
0.035 in.
1 μL
Inoculating Loop and Needle, Rigid
40 Pk. per Cs.