Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
Fisherbrand™ Magnetic Stirring Bars
Stirbar assortments come in a convenient storage box. Fisherbrand™ Magnetic Stirring Bars contain PTFE encapsulated Alnico magnets and come in various sizes.
 Afficher plus de versions de ce produit
Numéro de catalogue. 14-512-132
Découvrez plus d'offres spéciales
Description
Description
Stirbar assortments come in a convenient storage box. Fisherbrand™ Magnetic Stirring Bars contain PTFE encapsulated Alnico magnets and come in various sizes.
  • Choose from Octagon, Cylindrical, Polygon, Micro, Egg-Shaped or Spinplus™ shaped assortments
  • FDA Grade, USP Class VI PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclavable
Spécifications
Spécifications
One each in yellow, red, and blue, in the following sizes 0.5 x 0.13 in., 0.31 x 0.31 in., 0.88 x 0.31 in., 0.5 x 0.31 in., 1 x 0.31 in., 1.5 x 0.31 in., 2 x 0.31 in., and 3 x 0.5 in.
Stir Bar
Alnico Magnet
Octagon
PTFE
Yellow, Red, Blue
ISO 9001:2008
Videos
FDS
Documents
Documents
Accessoires
Offres spéciales
Offres spéciales