Fisherbrand™ Magnetic Stirring Bars
Stirbar assortments come in a convenient storage box. Fisherbrand™ Magnetic Stirring Bars contain PTFE encapsulated Alnico magnets and come in various sizes.
 View more versions of this product
Catalog No. 14-512-130
Explore available promotions
Description
Description
Stirbar assortments come in a convenient storage box. Fisherbrand™ Magnetic Stirring Bars contain PTFE encapsulated Alnico magnets and come in various sizes.
  • Choose from Octagon, Cylindrical, Polygon, Micro, Egg-Shaped or Spinplus™ shaped assortments
  • FDA Grade, USP Class VI PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclavable
Specifications
Specifications
Two each of the following sizes 0.5 x 0.38 in., 0.5 x 0.31 in., 0.75 x 0.31 in., 1 x 0.31 in., 1 x 0.38 in., 1.5 x 0.31 in., 1.75 x 0.31 in., 2 x 0.38 in., and 2 x 0.31 in., one each of the following 1.25 x 0.31 in., 1.25 x 0.38 in., 1.5 x 0.38 in., and 1
Stir Bar
Alnico Magnet
ISO 9001:2008
PTFE
White
Autoclavable
Sphere
Videos
SDS
Documents
Documents
Accessories