missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Micro Blood Collecting Tubes

With tapered bottom tips to facilitate micro blood sample collection

$125.31 - $140.72

Specifications

Product Type Micro Blood Collecting Tube
Material Soda Lime Glass
Sterility Non-sterile
Closure Type Tube Sealant
For Use With (Application) Natelson and Caraway techniques
View More Specs

Products 2
Catalog Number Mfr. No. Additive Closure Color Capacity (Metric) Length (Metric) Price Quantity  
Catalog Number Mfr. No. Additive Closure Color Capacity (Metric) Length (Metric) Price Quantity  
02-668-25
Accessories View Documents
N/A Ammonium Heparin Red 350 μL 75 mm
Pack of 100 Each for $140.72
Case of 6 Pack for $844.32
 
02-668-30
Accessories View Documents
N/A No Additive Blue 350 μL 75 mm
Pack of 100 Each for $125.31
Case of 6 Pack for $751.86
 
Description

Description

With tapered bottom tips to facilitate micro blood sample collection

  • For Natelson and Caraway techniques
  • Color-coded tops distinguish between plain and heparinized tubes
  • Heparinized tubes are treated with ammonium heparin
  • Natelson tubes have 1.44 to 1.76mm I.D., 147mm length
  • Caraway tubes have 2.59 to 2.81mm I.D., 75mm length

Specifications

Specifications

Micro Blood Collecting Tube
Non-sterile
Natelson and Caraway techniques
Soda Lime Glass
Tube Sealant
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'