Fisherbrand™ Micro Stir Bars
Micro Color-Coded Stirring Bar For stirring test tubes, micro-beakers, etc.
 View more versions of this product
Catalog No. 1451178
Explore available promotions
Description
Description
Micro Color-Coded Stirring Bar For stirring test tubes, micro-beakers, etc.
  • Octagonal shape
  • Resists corrosion, heat, and cold
  • Available in blue, red or yellow
  • PTFE
Specifications
Specifications
all sizes
PTFE
Multiple sizes
Yellow, Blue, Red
24
All Sizes
all sizes
Multiple sizes
Stir Bar
all sizes
PTFE
Octagon