Fisherbrand™ Micro Stir Bars
Micro Color-Coded Stirring Bar For stirring test tubes, micro-beakers, etc.
 View more versions of this product
Catalog No. 1451179
Explore available promotions
Description
Description
Micro Color-Coded Stirring Bar For stirring test tubes, micro-beakers, etc.
  • Octagonal shape
  • Resists corrosion, heat, and cold
  • Available in blue, red or yellow
  • PTFE
Specifications
Specifications
0.31 in.
Stir Bar Micro Red 8x1.5mm
Stir Bar
12
0.07 in.
0.06 in.
Autoclavable
Octagon
PTFE
0.31 x 0.07 in. (8 x 1.7mm)
Red
PTFE
1.5 mm
1.7mm
8 mm