Fisherbrand™ Micro Stir Bars
Micro Color-Coded Stirring Bar For stirring test tubes, micro-beakers, etc.
 View more versions of this product
Catalog No. 1451183
Explore available promotions
Description
Description
Micro Color-Coded Stirring Bar For stirring test tubes, micro-beakers, etc.
  • Octagonal shape
  • Resists corrosion, heat, and cold
  • Available in blue, red or yellow
  • PTFE
Specifications
Specifications
0.19 in.
Stir Bar Micro Yellow 5x2mm
Stir Bar
12
0.08 in.
0.07 in.
Autoclavable
Octagon
PTFE
0.19 x 0.08 in. (5 x 2mm)
Yellow
PTFE
2 mm
2mm
5 mm