Fisherbrand™ Micro Stir Bars
Micro Color-Coded Stirring Bar For stirring test tubes, micro-beakers, etc.
 View more versions of this product
Catalog No. 1451192
Explore available promotions
Description
Description
Micro Color-Coded Stirring Bar For stirring test tubes, micro-beakers, etc.
  • Octagonal shape
  • Resists corrosion, heat, and cold
  • Available in blue, red or yellow
  • PTFE
Specifications
Specifications
0.39 in.
Stir Bar Micro Yellow 10x3mm
Stir Bar, Micro
12
0.13 in.
0.118 in.
Autoclavable
Octagon
PTFE
0.39 x 0.13 in. (10 x 3.3mm)
Yellow
PTFE
3 mm
3.3mm
10 mm
Product Suggestions
Product Suggestions