Fisherbrand™ Micro Stir Bars
Micro Color-Coded Stirring Bar For stirring test tubes, micro-beakers, etc.
 View more versions of this product
Catalog No. 1451195
Explore available promotions
Description
Description
Micro Color-Coded Stirring Bar For stirring test tubes, micro-beakers, etc.
  • Octagonal shape
  • Resists corrosion, heat, and cold
  • Available in blue, red or yellow
  • PTFE
Specifications
Specifications
0.39 in.
Stir Bar Micro Blue 10x3mm
Stir Bar
12
0.126 in.
0.118 in.
Autoclavable
Octagon
PTFE
0.39 x 0.126 in. (10 x 3.2mm)
Blue
PTFE
3 mm
3.2mm
10 mm