Fisherbrand™ Micro Stir Bars
Micro Color-Coded Stirring Bar For stirring test tubes, micro-beakers, etc.
 View more versions of this product
Catalog No. 1451199
Explore available promotions
Description
Description
Micro Color-Coded Stirring Bar For stirring test tubes, micro-beakers, etc.
  • Octagonal shape
  • Resists corrosion, heat, and cold
  • Available in blue, red or yellow
  • PTFE
Specifications
Specifications
0.5 in.
Stir Bar Micro Blue 12.7x3mm
Stir Bar
12
0.124 in.
0.118 in.
Autoclavable
Octagon
PTFE
0.5 x 0.124 in. (12.7 x 3.14mm)
Blue
PTFE
3 mm
3.14mm
12.7 mm