Fisherbrand™ Microcentrifuge Tubes with Tether Cap
Reduce sample contamination by keeping cap with tube
 View more versions of this product
Catalog No. 02-707-354
Explore available promotions
Description
Description
Reduce sample contamination by keeping cap with tube
Natural-colored polypropylene tubes include leash-and-loop tether. Ultragrade polypropylene caps are fitted with ethylene/propylene O-rings. Available sterile or nonsterile with round or skirted bottoms.
  • Removable tether rotates freely for easy opening and closing
  • O-ring forms positive seal with tube to reduce leakage
  • Withstand 20,000xG
  • Suitable for vapor-phase liquid nitrogen storage, boiling and autoclaving
  • RNase and DNase free
Order Info
Nonsterile tubes come separate from tether caps; sterile tubes come attached; 500 per pack
Specifications
Specifications
Polypropylene
2 mL
Polypropylene
Bag
Tether Cap
20,000 x g
Autoclavable
Natural
Suitable for vapor-phase liquid nitrogen storage, boiling and autoclaving
Non-sterile
Round
Videos
SDS
Documents
Documents
Accessories