Fisherbrand™ Nonsterile Threaded End Microcentrifuge Tubes without Caps
Choose skirted- or conical-bottom style.
 View more versions of this product
Catalog No. 02-681-334
Description
Description
Choose skirted- or conical-bottom style.
  • Threaded to accept colored screw caps (sold separately)
  • Siliconized/low-retention conical-bottom tubes also available
Note: For caps, see Fisherbrand™ Colored Screw Caps. 
Specifications
Specifications
25,000 x g
Natural
Non-sterile
Screw Cap
0.5 mL
Plastic
No
Conical
SDS
Documents
Documents
Accessories