Fisherbrand™ Nonsterile Threaded End Microcentrifuge Tubes without Caps
Choose skirted- or conical-bottom style.
 View more versions of this product
Catalog No. 02-681-340
Explore available promotions
Description
Description
Choose skirted- or conical-bottom style.
  • Threaded to accept colored screw caps (sold separately)
  • Siliconized/low-retention conical-bottom tubes also available
Note: For caps, see Fisherbrand™ Colored Screw Caps. 
Specifications
Specifications
30,000 x g
Amber
Non-sterile
Conical
1.5 mL
Plastic
Screw Cap
Videos
SDS
Documents
Documents
Accessories