Fisherbrand™ Nonsterile Threaded End Microcentrifuge Tubes without Caps
Choose skirted- or conical-bottom style.
Description
Choose skirted- or conical-bottom style.
- Threaded to accept colored screw caps (sold separately)
- Siliconized/low-retention conical-bottom tubes also available
Specifications
18,000 x g | |
Amber | |
Non-sterile | |
Skirted |
2 mL | |
Plastic | |
Screw Cap |