Fisherbrand™ Oil-Selective Sorbents
Selectively remove hydrocarbons from water
$180.14 - $180.14
Specifications
Material | Polypropylene |
---|---|
For Use With (Application) | Sorbing oil and a wide variety of acids and other hazardous chemicals |
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Type | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Type | Price | Quantity | |||
17-215-2A
|
Fisherbrand™
FBB810 |
70 gal. | 265 L | Oil Sorbent Boom |
Pack of 4 Each for $180.14
|
|
|||
Description
Multilayer fine fiber polypropylene sorbents sorb up to 25 times their weight in petrochemical-based liquids.
- For sorbing oil and a wide variety of acids and other hazardous chemicals
- Unaffected by temperature, humidity and pest infestation; easy to store
- Can be cleaned and reused repeatedly
Booms
- Polypropylene sorbents absorb oil and solvent spills on water
- Booms available with Sock-in-Net or open netting covering designs
- Booms float on the surface to selectively remove hydrocarbons and contain the spill
- Hardware is included to link booms
- Multilayer fine fiber material for cleaning spills of flammable material
- Sorbent pads and rolls come in a range of sizes and can be trimmed to fit
- Compact pillow, socks, and pads can be placed directly on spills
- Can be scattered on petroleum spills
- Use particulates exactly where needed
Specifications
Polypropylene | |
Sorbing oil and a wide variety of acids and other hazardous chemicals |