missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Autoclavable Biohazard Waste Bags, with Indicator
Printed in English and Spanish.
Supplier: Fisherbrand™ 1814B
Description
Printed in English and Spanish
- Maximum temperature of 140°C (285°F); use at 125°C (257°F)
- Come with wire-tie closures
- With sterilization indicator
Specifications
ASTM D1709, Dart impact | |
Orange | |
58.4 cm | |
Autoclavable | |
23 in. | |
Biohazard Waste Bags with indicator |
Wire Tie | |
23 x 19 in. (58.4 x 48.2 cm) | |
48.2 cm | |
19 in. | |
Polypropylene | |
140°C |