missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Autoclavable Biohazard Waste Bags, with Indicator

Printed in English and Spanish.

Supplier:  Fisherbrand™ 18151

 View more versions of this product

Catalog No. 01-815-1



Explore available promotions

Description

Description

Printed in English and Spanish

 

  • Maximum temperature of 140°C (285°F); use at 125°C (257°F)
  • Come with wire-tie closures
  • With sterilization indicator

 

Specifications

Specifications

ASTM D1709, Dart impact
Orange
28 cm
Autoclavable
11 in.
Biohazard Waste Bags with indicator
Wire Tie
11 x 8.5 in. (28 x 21.5 cm)
21.5 cm
8.5 in.
Polypropylene
140°C
Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions