missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Autoclavable HD Biohazard Waste Bags

Biohazard symbol printed bags to ensure safe handling and disposal of biohazardous waste.

Supplier:  Fisherbrand™ 1815C

 View more versions of this product

Catalog No. 01-815C


Explore available promotions

Description

Description

Impact- and tear-resistant autoclave bags for waste sterilization. Fisherbrand™ Orange Autoclave Bags without Sterilization Indicator are constructed of high-density polyethylene.

  • Impact- and tear-resistant.
  • Bright orange color.
  • High-density polyethylene construction

Compliance

Pass ASTM™ impact- and tear-resistance tests.

Specifications

Specifications

ASTM
Orange with Black Print
88.9 cm
Autoclavable
24 in.
38 micron
127°C
None
35 x 24 in. (88.9 x 61 cm)
61 cm
Printed in English
35 in.
HDPE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions