Fisherbrand™ Paper pH Strips
An easy choice for classroom experiments or demonstrations
Supplier:  Fisherbrand™ B50-SHTRG-100120-SST
 View more versions of this product
Catalog No. 13-640-513
Explore available promotions
Description
Description
  • Ideal for educational use
  • Low-cost paper strips are easy to pass around the classroom
  • Easy-to-use—simply dip strip in solution to be tested and match to color chart immediately
Order Info
Box of 10 vials, 100 low-cost paper strips per vial.
Specifications
Specifications
10 to 12
0.5
Room Temperature
For classroom experiments or demonstrations
100 Tests/Vial, 10 Vials/Cs.
pH Test Paper