Fisherbrand™ Paper pH Strips
An easy choice for classroom experiments or demonstrations
Supplier:  Fisherbrand™ B50-SHTRG-000140-SET
 View more versions of this product
Catalog No. 13-640-515
Explore available promotions
Description
Description
  • Ideal for educational use
  • Low-cost paper strips are easy to pass around the classroom
  • Easy-to-use—simply dip strip in solution to be tested and match to color chart immediately
Order Info
Box of 10 vials, 100 low-cost paper strips per vial.
Specifications
Specifications
0 to 14
1 vial each of 13-640-511, 13-640-509, 13-640-512, 13-640-513, and 13-640-514. 2 vials of 13-640-510, 3 vials of 13-640-508
Room Temperature
For classroom experiments or demonstrations
100 Tests/Vial, 10 Vials/Cs.
pH Test Paper Set