Fisherbrand™ Petri Dishes Specialty
Multiple sizes and styles for variety of applications
 View more versions of this product
Catalog No. FB087579B
Explore available promotions
Description
Description
Multiple sizes and styles for variety of applications
  • All dishes manufactured from virgin medical grade polystyrene
  • All dishes sterilized using e-beam sterilization
  • Deep style petri dishes available - designed for increased media volumes, allowing for longer culture times
  • Contact plates available for collection in environments where contact is required to collect the sample
  • Convenient packaging in easy open sleeves
  • 25 Sleeves/Cs.
Specifications
Specifications
Slippable
Polystyrene
Sterile
20/Sleeve, 25 Sleeves/Cs.
15 mm
Flat
Round
Mono-Plate
100 x 15mm
100 mm