Fisherbrand™ Pipet Holder
Fisherbrand Pipet Sterilizing/Drying Racks are perfect for holding pipets during cleansing and sterilizing
 View more versions of this product
Catalog No. 13-814-52
Explore available promotions
Description
Description
Available in two convenient sizes: 2.5 x 13.25 in (6.4 x 33.7 cm) and 4.75 x 13.25 in.(12.1 x 33.7 cm)
  • Inclined trough shape at 33° angle for cleansing, drying and storage of pipets
  • Riveted aluminum with smooth corners and edges
  • Can withstand indefinite hot air or steam sterilization
  • Made from ASTM standard (899-94) stainless steel
Specifications
Specifications
Not Autoclavable
4.75 in.
13.25 in.
12.1 cm
33.7 cm