Fisherbrand™ Pipet Tips in SureGrip Racks
Tips in sturdy, reusable racks
 View more versions of this product
Catalog No. 02-100-509
Explore available promotions
Description
Description
Pipet Tips in Racks
  • All tips are available racked in sturdy, reusable CellRacks
  • Racks can withstand numerous autoclave cycles
  • Keeps tips aligned for pipetter mounting
Several Tip Sizes Available
  • 10μL Pipet tip features graduation marks at 2μL and 10μL
  • 10μL Long Reach tip features extended length (45mm) that reaches the bottom of tubes without contaminating the pipetter shaft, and graduation marks at 2μL, 5μL and 10μL
  • 200μL Pipet tip features a soft fit collar for easy mounting and eject, graduation marks at 10μL, 50μL and 100μL, and is available in natural and yellow
  • 1250μL Pipet tips feature an extended length (103mm), racked on 9mm centers (multichannel compatible), and graduation marks at 200μL, 500μL and 1000μL
Specifications
Specifications
200 μL
Yellow
Rack
Yes
Sterile
Autoclavable
Extended Length