Fisherbrand™ Plastic Joint Clips
Made of plastic for use on standard taper glass joints
Supplier:  Fisherbrand™ 14955252
 View more versions of this product
Catalog No. 14-955-252
Explore available promotions
Description
Description
Made of plastic for use on standard taper glass joints
  • Color coded for easy size identification
  • Does not corrode
  • Will not scratch glass
  • Withstands temperatures up to 150°C
Specifications
Specifications
10/Pk.
Plastic
Purple
Joint Clip
Standard taper Glass joint size 12