Fisherbrand™ Plastic Joint Clips
Made of plastic for use on standard taper glass joints
Supplier: Fisherbrand™ 14955253
Description
Made of plastic for use on standard taper glass joints
- Color coded for easy size identification
- Does not corrode
- Will not scratch glass
- Withstands temperatures up to 150°C
Specifications
10/Pk. | |
Plastic | |
Yellow |
Joint Clip | |
Standard taper Glass joint size 14 |