Fisherbrand™ Plastic Joint Clips
![Edge](/content/dam/fishersci/glyphs/edge.png)
Made of plastic for use on standard taper glass joints
Supplier: Fisherbrand™ 14955255
Description
Made of plastic for use on standard taper glass joints
- Color coded for easy size identification
- Does not corrode
- Will not scratch glass
- Withstands temperatures up to 150°C
Specifications
10/Pk. | |
Plastic | |
Green |
Joint Clip | |
Standard taper Glass joint size 24 |