Fisherbrand™ Plastic pH Strips
Laboratory grade plastic pH strips in durable and easy to open flip-top vials
Supplier: Fisherbrand™ B60-SHTRG-065130-VPS
Description
Laboratory grade plastic pH strips in durable and easy to open flip-top vials
- Easy-to-use — simply dip strip in solution to be tested and match to color chart immediately
- Single color match with full 0.5 in. (1.27cm) matching area for quick and accurate matches
- Flip-top vial stays closed when dropped, but is easy to open with one-hand
- Sturdy vial keeps strips safe from the elements
Order Info
Box 6 Vials, 100 strips per vial.
Specifications
6.5 to 13 | |
For quick and accurate matches | |
100 Strips/Vial, 6 Vials/Pk. | |
pH Strip |
Strip | |
0.5 | |
Room Temperature |