Fisherbrand™ Plastic pH Strips Edge
Edge
Laboratory grade plastic pH strips in durable and easy to open flip-top vials
Supplier:  Fisherbrand™ B60-SHTRG-050090-VPS
 View more versions of this product
Catalog No. 13-640-519
Edge
Explore available promotions
Description
Description
Laboratory grade plastic pH strips in durable and easy to open flip-top vials
  • Easy-to-use — simply dip strip in solution to be tested and match to color chart immediately
  • Single color match with full 0.5 in. (1.27cm) matching area for quick and accurate matches
  • Flip-top vial stays closed when dropped, but is easy to open with one-hand
  • Sturdy vial keeps strips safe from the elements
Order Info
Box 6 Vials, 100 strips per vial.
Specifications
Specifications
5 to 9
For quick and accurate matches
100 Strips/Vial, 6 Vials/Pk.
pH Strip
Strip
0.5
Room Temperature