Fisherbrand™ Polyethylene Bags
Flexible even when frozen
 View more versions of this product
Catalog No. 19075388A
Explore available promotions
Description
Description
Flexible even when frozen
  • Ideal for short-term storage
  • May be tied or heat-sealed
  • Not suggested for use with liquids
  • Thickness, 4mil
Specifications
Specifications
Heat Seal or Tie
4 mil
12 in.
8 in.
Polyethylene
12 x 8 in. (30.5 x 20.3cm)
30.5 cm
20.3 cm