Fisherbrand™ Polyethylene Bags
Flexible even when frozen
 View more versions of this product
Catalog No. 19-075-388B
Explore available promotions
Description
Description
Flexible even when frozen
  • Ideal for short-term storage
  • May be tied or heat-sealed
  • Not suggested for use with liquids
  • Thickness, 4mil
Specifications
Specifications
Heat Seal or Tie
4 mil
10 in.
6 in.
Polyethylene
10 x 6 in. (25.4 x 15.2cm)
25.4 cm
15.2 cm