Fisherbrand™ Polyethylene Bags
Flexible even when frozen
 View more versions of this product
Catalog No. 19-075-388C
Explore available promotions
Description
Description
Flexible even when frozen
  • Ideal for short-term storage
  • May be tied or heat-sealed
  • Not suggested for use with liquids
  • Thickness, 4mil
Specifications
Specifications
Heat Seal or Tie
4 mil
16 in.
10 in.
Polyethylene
16 x 10 in. (40.6 x 25.4cm)
40.6 cm
25.4 cm