Description
Flexible even when frozen
- Ideal for short-term storage
- May be tied or heat-sealed
- Not suggested for use with liquids
- Thickness, 4mil
Specifications
Polyethylene | |
18 x 12 in. (45.7 x 30.5cm) | |
45.7 cm | |
30.5 cm |
4 mil | |
18 in. | |
12 in. |