Description
Flexible even when frozen
- Ideal for short-term storage
- May be tied or heat-sealed
- Not suggested for use with liquids
- Thickness, 4mil
Specifications
Polyethylene | |
12 x 10 in. (30.5 x 25.4cm) | |
30.5 cm | |
25.4 cm |
4 mil | |
12 in. | |
10 in. |