Fisherbrand™ Polyethylene Bags
Flexible even when frozen
 View more versions of this product
Catalog No. 19-075-388K
Explore available promotions
Description
Description
Flexible even when frozen
  • Ideal for short-term storage
  • May be tied or heat-sealed
  • Not suggested for use with liquids
  • Thickness, 4mil
Specifications
Specifications
Heat Seal or Tie
4 mil
20 in.
12 in.
Polyethylene
20 x 12 in. (50.8 x 30.5cm)
50.8 cm
30.5 cm