missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Autoclavable HD Biohazard Waste Bags

Autoclavable, polyethylene biohazard bags ensure safe handling and disposal of biohazardous waste.

Supplier:  Fisherbrand™ 18143

 View more versions of this product

Catalog No. 01-814-3


Explore available promotions

Description

Description

Autoclavable, polyethylene biohazard bags to ensure safe handling and disposal of biohazardous waste.

  • All bags supplied with autoclavable elastic steam-venting closures except 1.5 mil-thick plain clear bags
  • Polyethylene
  • Temperature range 260°F
  • Available in 4 colors/styles
Note: This bag (01-814-3) has no biohazard symbol printed on it, and no indicator.  

Specifications

Specifications

ASTM D1709-85
This bag has no biohazard symbol, and no indicator.
24 x 36 in. (61 x 91 cm)
61 cm
24 in.
1.50 mil
Autoclavable Bags
None
Natural
91 cm
Autoclavable
36 in.
Polyethylene
127°C
Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions