missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Autoclavable HD Biohazard Waste Bags

Autoclavable, polyethylene biohazard bags ensure safe handling and disposal of biohazardous waste.

Supplier:  Fisherbrand™ 1829F

 View more versions of this product

Catalog No. 01-829F


Explore available promotions

Description

Description

Autoclavable, polyethylene biohazard bags to ensure safe handling and disposal of biohazardous waste.

  • All bags supplied with autoclavable elastic steam-venting closures except 1.5 mil-thick plain clear bags
  • Polyethylene
  • Temperature range 260°F
  • Available in 4 colors/styles
Specifications

Specifications

OSHA, 29 CFR1910.1030
Red with Black Print
76 cm
Autoclavable
33 in.
1.50 mil
Autoclavable Bags
Elastic
30 x 33 in. (76 x 84 cm)
84 cm
Printed in English and Spanish
30 in.
Polyethylene
127°C
Videos
SDS
Documents

Documents

Promotions

Promotions