Learn More
Fisherbrand™ Polystyrene Beakers
Ideal for use as sample containers
Description
Ideal for use as sample containers
- 20mL size is useful in flame photometry
- 5mL size can be used in atomic absorption spectroscopy and flame photometry
Specifications
| 10 mL | |
| Polystyrene | |
| Beaker | |
| Single-use |