Fisherbrand™ Powder Funnels
Parallel stems minimize bridging of powder.
Supplier:  Fisherbrand™ 3007000065
 View more versions of this product
Catalog No. 10-500
Explore available promotions
Description
Description
Parallel stems minimize bridging of powder
  • Durable polypropylene
  • External ribbing prevents air locks
  • Autoclavable
Specifications
Specifications
6.5cm
16mm
6.6 cm
22mm
50 mL
Polypropylene
Autoclavable